Learn More
ATCC Escherichia coli; Strain TY-2482; O104:H4
Catalog No. 502385007
Encompass
Escherichia coli strain TY-2482 was isolated in 2011 from a stool sample a from patient with hemolytic uremic syndrome (HUS). This bacterium is serotype O104:H4 and is known to carry the aggR and stx2 genes.
Catalog No. 50-238-5007 Supplier ATCC Supplier No. BAA2326
Escherichia coli strain TY-2482 was isolated in 2011 from a stool sample a from patient with hemolytic uremic syndrome (HUS). This bacterium is serotype O104:H4 and is known to carry the aggR and stx2 genes.