Learn More
ATCC Escherichia coli; W1485
Catalog No. 502383323
Encompass
Escherichia coli strain W1485 is a lambda-derivative of E. coli strain K-12. It is an indicator strain that is lysed by lambda bacteriophage.
Catalog No. 50-238-3323 Supplier ATCC Supplier No. 12435
Escherichia coli strain W1485 is a lambda-derivative of E. coli strain K-12. It is an indicator strain that is lysed by lambda bacteriophage.