Learn More
ATCC Escherichia coli; W3110
Catalog No. 502382064
Encompass
Escherichia coli strain W3110 is a whole-genome sequenced bacterium that can be used as a transformation host. This strain is a prototrophic derivative of strain K-12.
Catalog No. 50-238-2064 Supplier ATCC Supplier No. 27325
Escherichia coli strain W3110 is a whole-genome sequenced bacterium that can be used as a transformation host. This strain is a prototrophic derivative of strain K-12.