Learn More
Medchemexpress LLC Escin | 6805-41-0 | 96.6% | 250 MG
SDP
Catalog No. 5000414342
Encompass_Preferred
Escin is a natural compound of triterpenoid saponins isolated from horse chestnut (Aesculus hippocastanum) seeds. It acts as a vasoprotective anti-inflammatory, anti-edematous, and anti-nociceptive agent. It is for research use only and not sold to patients.
  • Used as a vasoprotective anti-inflammatory agent
  • Acts as an anti-edematous agent
  • Functions as an anti-nociceptive agent
  • Isolated from horse chestnut seeds
Catalog No. 50-004-14342 Supplier Medchemexpress LLC Supplier No. 250MGHYB2114
Escin is a natural compound of triterpenoid saponins isolated from horse chestnut (Aesculus hippocastanum) seeds. It acts as a vasoprotective anti-inflammatory, anti-edematous, and anti-nociceptive agent. It is for research use only and not sold to patients.
  • Used as a vasoprotective anti-inflammatory agent
  • Acts as an anti-edematous agent
  • Functions as an anti-nociceptive agent
  • Isolated from horse chestnut seeds