Learn More
Medchemexpress LLC HY-N0554 5mg Medchemexpress, Escin IA CAS:123748-68-5 Purity:>98%
SDP
Catalog No. 501970285
Encompass_Preferred
Medchemexpress, HY-N0554 5mg Escin IA CAS:123748-68-5 Escin IA is a triterpene saponin isolated from horse chestnut, which inhibits HIV-1 protease with IC50 values of 35 μM. Escin IA has anti-TNBC metastasis activity, and its action mechanisms involved inhibition of epithelial-mesenchymal transition process by down-regulating LOXL2 expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0285 Supplier Medchemexpress LLC Supplier No. HYN05545MG
Medchemexpress, HY-N0554 5mg Escin IA CAS:123748-68-5 Escin IA is a triterpene saponin isolated from horse chestnut, which inhibits HIV-1 protease with IC50 values of 35 μM. Escin IA has anti-TNBC metastasis activity, and its action mechanisms involved inhibition of epithelial-mesenchymal transition process by down-regulating LOXL2 expression. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.