Learn More
Medchemexpress LLC Escin IA (standard) | 123748-68-5 | 1131.26 g/mol | C55H86O24 | 10 MG
SDP
Catalog No. 5000295653
Encompass_Preferred
Escin IA is a triterpene saponin isolated from Aesculus hippocastanum and provided as an analytical standard for research applications. It inhibits HIV-1 protease (IC50 ≈ 35 μM) and has demonstrated anti-metastatic activity in triple-negative breast cancer models by inhibiting epithelial-mesenchymal transition and down-regulating LOXL2 expression.
  • Triterpene saponin natural product
  • Inhibits HIV-1 protease (IC50 ~35 μM)
  • Exhibits anti-TNBC metastasis activity via EMT inhibition
  • Intended for research and analytical applications
  • Available as a solid, common pack size 10 MG
  • Molecular weight 1131.26 g/mol; CAS 123748-68-5
Catalog No. 50-002-95653 Supplier Medchemexpress LLC Supplier No. HYN055410MG
Escin IA is a triterpene saponin isolated from Aesculus hippocastanum and provided as an analytical standard for research applications. It inhibits HIV-1 protease (IC50 ≈ 35 μM) and has demonstrated anti-metastatic activity in triple-negative breast cancer models by inhibiting epithelial-mesenchymal transition and down-regulating LOXL2 expression.
  • Triterpene saponin natural product
  • Inhibits HIV-1 protease (IC50 ~35 μM)
  • Exhibits anti-TNBC metastasis activity via EMT inhibition
  • Intended for research and analytical applications
  • Available as a solid, common pack size 10 MG
  • Molecular weight 1131.26 g/mol; CAS 123748-68-5