Learn More
Medchemexpress LLC HY-N0555 5mg Medchemexpress, Escin IB CAS:26339-90-2 Purity:>98%
SDP
Catalog No. 501970286
Encompass_Preferred
Medchemexpress, HY-N0555 5mg Escin IB CAS:26339-90-2 Escin IB is a saponin isolated from skin and the endosperm of seeds of horse chestnut (Aesculus hippocastanum). Escin IB shows inhibitory effect on pancreatic lipase activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0286 Supplier Medchemexpress LLC Supplier No. HYN05555MG
Medchemexpress, HY-N0555 5mg Escin IB CAS:26339-90-2 Escin IB is a saponin isolated from skin and the endosperm of seeds of horse chestnut (Aesculus hippocastanum). Escin IB shows inhibitory effect on pancreatic lipase activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.