Learn More
Medchemexpress LLC Escitalopram | 128196-01-0 | MFCD09817397 | >98.0% | C20H21FN2O | 10MG
SDP
Catalog No. 5000261019
Encompass_Preferred
Escitalopram is the S-enantiomer of racemic citalopram and a selective serotonin reuptake inhibitor (SSRI). This compound is provided as a research/analytical standard for preclinical pharmacology, assay development, and binding studies of serotonin transporter activity.
  • High selectivity for the serotonin transporter, enabling focused pharmacological studies.
  • Well-characterized chemical identity with CAS 128196-01-0 and formula C20H21FN2O.
  • Suitable for analytical standards, assay development, and quantitative studies.
  • Available in small analytical pack sizes appropriate for research use.
  • Useful for structure-activity and binding affinity experiments.
Catalog No. 50-002-61019 Supplier Medchemexpress LLC Supplier No. HY1425810MG
Escitalopram is the S-enantiomer of racemic citalopram and a selective serotonin reuptake inhibitor (SSRI). This compound is provided as a research/analytical standard for preclinical pharmacology, assay development, and binding studies of serotonin transporter activity.
  • High selectivity for the serotonin transporter, enabling focused pharmacological studies.
  • Well-characterized chemical identity with CAS 128196-01-0 and formula C20H21FN2O.
  • Suitable for analytical standards, assay development, and quantitative studies.
  • Available in small analytical pack sizes appropriate for research use.
  • Useful for structure-activity and binding affinity experiments.