Learn More
Medchemexpress LLC Escitalopram-d6 oxalate | 1217733-09-9 | C22H17D6FN2O5 | 1 MG
SDP
Catalog No. 5000353659
Encompass_Preferred
Escitalopram-d6 oxalate is a deuterium-labeled form of Escitalopram oxalate, a selective serotonin reuptake inhibitor (SSRI) with high binding affinity and selectivity. It is an antidepressant used in research for major depression. Deuterium-labeled compounds are valuable as tracers and internal standards in drug development and quantitative analysis.
  • Deuterium labeled compound
  • Functions as a selective serotonin reuptake inhibitor (SSRI)
  • Applicable as a tracer in research
  • Suitable as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Used for major depression research
  • Contains stable heavy isotopes for drug development quantitation
Catalog No. 50-003-53659 Supplier Medchemexpress LLC Supplier No. HY14258AS1MG
Escitalopram-d6 oxalate is a deuterium-labeled form of Escitalopram oxalate, a selective serotonin reuptake inhibitor (SSRI) with high binding affinity and selectivity. It is an antidepressant used in research for major depression. Deuterium-labeled compounds are valuable as tracers and internal standards in drug development and quantitative analysis.
  • Deuterium labeled compound
  • Functions as a selective serotonin reuptake inhibitor (SSRI)
  • Applicable as a tracer in research
  • Suitable as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Used for major depression research
  • Contains stable heavy isotopes for drug development quantitation