Learn More
Selleck Chemical LLC Escitalopram Oxalate S4064-50mg
SDP
Catalog No. 503109651
Encompass_Preferred
Escitalopram Oxalate is a selective serotonin (5-HT) reuptake inhibitor (SSRI) with Ki of 0 89 nM
Catalog No. 50-310-9651 Supplier Selleck Chemical LLC Supplier No. S406450MG
Escitalopram Oxalate is a selective serotonin (5-HT) reuptake inhibitor (SSRI) with Ki of 0 89 nM