Learn More
Medchemexpress LLC Escitalopram oxalate | 219861-08-2 | 100.0% | C22H23FN2O5 | 50 MG
SDP
Catalog No. 5000353577
Encompass_Preferred
Escitalopram oxalate is an analytical standard for Escitalopram (oxalate). This S-enantiomer of racemic Citalopram functions as a selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM and is used in research for major depression. It is intended for research and analytical applications.
  • Exhibits approximately 30-fold higher binding affinity than its R(-)-enantiomer
  • Shows selectivity over both dopamine transporter (DAT) and norepinephrine transporter (NET)
  • Commonly utilized in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS
Catalog No. 50-003-53577 Supplier Medchemexpress LLC Supplier No. HY14258AR50MG
Escitalopram oxalate is an analytical standard for Escitalopram (oxalate). This S-enantiomer of racemic Citalopram functions as a selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM and is used in research for major depression. It is intended for research and analytical applications.
  • Exhibits approximately 30-fold higher binding affinity than its R(-)-enantiomer
  • Shows selectivity over both dopamine transporter (DAT) and norepinephrine transporter (NET)
  • Commonly utilized in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS