Learn More
Medchemexpress LLC Escitalopram oxalate | 219861-08-2 | 99.5% | C₂₂H₂₃FN₂O₅ | 25 MG
SDP
Catalog No. 5000356716
Encompass_Preferred
Escitalopram oxalate, the S-enantiomer of racemic citalopram, is a selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM. It demonstrates approximately 30-fold higher binding affinity than its R(-)-enantiomer and exhibits selectivity over both dopamine transporter (DAT) and norepinephrine transporter (NET). This compound is an antidepressant used in research for major depression.
  • Selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM.
  • Demonstrates approximately 30-fold higher binding affinity than its R(-)-enantiomer.
  • Exhibits selectivity over both dopamine transporter (DAT) and norepinephrine transporter (NET).
  • Used in research for major depression.
  • Store at 4°C, sealed and dry, away from light and moisture.
Catalog No. 50-003-56716 Supplier Medchemexpress LLC Supplier No. 25MGHY14258AR
Escitalopram oxalate, the S-enantiomer of racemic citalopram, is a selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM. It demonstrates approximately 30-fold higher binding affinity than its R(-)-enantiomer and exhibits selectivity over both dopamine transporter (DAT) and norepinephrine transporter (NET). This compound is an antidepressant used in research for major depression.
  • Selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM.
  • Demonstrates approximately 30-fold higher binding affinity than its R(-)-enantiomer.
  • Exhibits selectivity over both dopamine transporter (DAT) and norepinephrine transporter (NET).
  • Used in research for major depression.
  • Store at 4°C, sealed and dry, away from light and moisture.