Learn More
Apexbio Technology LLC Escitalopram Oxalate 219861-08-2 50mg
SDP
Catalog No. 502240779
Encompass_Preferred
Escitalopram Oxalate (CAS 219861-08-2) is the S-( )-enantiomer of citalopram and functions as a selective serotonin reuptake inhibitor (SSRI) It exhibits high affinity for the human serotonin transporter (5-HTT) inhibiting [3H]-5-HT uptake with a Ki of 6 6 nM in COS-1 cells expressing 5-HTT and displacing [125I]-RTI-55 with a Ki of 3 9 nM In rat brain synaptosomes Escitalopram Oxalate blocks the accumulation of [3H]-serotonin (IC50 2 1 nM) with markedly lower activity against norepinephrine and dopamine uptake It also shows in vitro affinity for rat histamine H1 receptors and 1 sites Escitalopram Oxalate is widely utilized in neuropharmacological research related to depression and anxiety mechanisms
This item is not returnable. View return policy
Catalog No. 50-224-0779 Supplier Apexbio Technology LLC Supplier No. B223850
Escitalopram Oxalate (CAS 219861-08-2) is the S-( )-enantiomer of citalopram and functions as a selective serotonin reuptake inhibitor (SSRI) It exhibits high affinity for the human serotonin transporter (5-HTT) inhibiting [3H]-5-HT uptake with a Ki of 6 6 nM in COS-1 cells expressing 5-HTT and displacing [125I]-RTI-55 with a Ki of 3 9 nM In rat brain synaptosomes Escitalopram Oxalate blocks the accumulation of [3H]-serotonin (IC50 2 1 nM) with markedly lower activity against norepinephrine and dopamine uptake It also shows in vitro affinity for rat histamine H1 receptors and 1 sites Escitalopram Oxalate is widely utilized in neuropharmacological research related to depression and anxiety mechanisms