Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Escort™ Elf Air Sampling Pump

Catalog No. 111003036 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Flow Rate:
0.5 to 3.0 L/min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
11-100-3036 0.5 to 3.0 L/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3036 Supplier MilliporeSigma™ Supelco™ Supplier No. 28160U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Internal secondary standard calibrates the pump continuously, maintaining OSHA compliance

  • Patented electronic laminar flow control
  • Check against a primary standard only once a month or every 200 hours for coal mine dust sampling
  • Inlet filter traps moisture and particular matter
  • Stainless steel-filled plastic case provides EMI/RFI protection
  • Rated IP66 for resistance to water and dust ingress
  • Flow-Fault Indicator shows any sample inlet blockage
  • Run/Hold feature allows time out during sampling process
  • Elapsed time readout and low-battery warning
  • Exceptionally quiet operation
  • Low flow and dual tube sampling available with Gemini Twin-Port Sampler (28118-U)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH Certified for Coal Mine Dust Sampling (TC-74-030), MSHA Certified as Intrinsically Safe for Underground Use (Approval # 2 G-3924-1), UL Approved as Intrinsically Safe for use in Hazardous Locations
Voltage 4.8 V
Dimensions (L x W x H) 2.25 in. x 4 in. x 4.25 in. (5.7 cm x 10.2 cm x 10.8 cm)
Electrical Requirements 4.8 V Battery Pk. of 4 NiMH Cells
Weight (English) 23 oz.
Product Type Air Sampling Pump
Flow Rate 0.5 to 3.0 L/min.
Weight (Metric) 652 g
Operating Temperature Range (English) 32° to 113°F
Operating Temperature Range (Metric) 0° to 45°C

Escort is a registered trademark of Zefon International

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.