Learn More
Sigma Aldrich Fine Chemicals Biosciences Escort IV Transfection Reagent, 1 mL
Catalog No. NC1359189
Encompass
Escort IV is a unique formulation of a proprietary polycationic lipid and a neutral non-transfecting lipid. This liposome-forming compound is used for transfection of nucleic acids into a wide variety of eukaryotic cell types.
Catalog No. NC1359189 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L32871ML
Add to Cart
Add to Cart
Escort IV is a unique formulation of a proprietary polycationic lipid and a neutral non-transfecting lipid. This liposome-forming compound is used for transfection of nucleic acids into a wide variety of eukaryotic cell types.