Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Esculentoside A | 65497-07-6 | MFCD27980582 | 99.9% | 826.96 | C42H66O16 | 10 MG
SDP

Catalog No. 5000296440
Encompass_Preferred

Esculentoside A is a triterpene saponin isolated from the roots of Phytolacca esculenta and supplied as a research-grade analytical reagent. It demonstrates anti-inflammatory activity through inhibition of cyclooxygenase-2 (COX-2) and modulation of NF-κB and MAPK signaling pathways, and is intended for in vitro and experimental studies.

  • High purity: 99.9% (manufacturer reported)
  • CAS number: 65497-07-6
  • Molecular weight: 826.96 g/mol
  • Suitable for inflammation and immunology research
  • Available in milligram quantities for analytical use

Catalog No. 50-002-96440 Supplier Medchemexpress LLC Supplier No. HYN063210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Esculentoside A is a triterpene saponin isolated from the roots of Phytolacca esculenta and supplied as a research-grade analytical reagent. It demonstrates anti-inflammatory activity through inhibition of cyclooxygenase-2 (COX-2) and modulation of NF-κB and MAPK signaling pathways, and is intended for in vitro and experimental studies.

  • High purity: 99.9% (manufacturer reported)
  • CAS number: 65497-07-6
  • Molecular weight: 826.96 g/mol
  • Suitable for inflammation and immunology research
  • Available in milligram quantities for analytical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.