Learn More
Medchemexpress LLC Esculentoside H | 66656-92-6 | 5 MG
SDP
Catalog No. 5000418227
Encompass_Preferred
Esculentoside H (EsH) is a saponin isolated from the root extract of the perennial plant *Phytolacca esculenta*. It exhibits anti-tumor activity and suppresses colon cancer cell migration by blocking the JNK1/2 and NF-κB signaling-mediated matrix metalloproteinases-9 (MMP-9) expression, also linked to its capacity for TNF release.
  • Saponin isolated from the root extract of *Phytolacca esculenta*.
  • Demonstrates anti-tumor activity.
  • Inhibits colon cancer cell migration.
  • Blocks JNK1/2 and NF-κB signaling.
  • Suppresses matrix metalloproteinases-9 (MMP-9) expression.
  • Linked to TNF release.
Catalog No. 50-004-18227 Supplier Medchemexpress LLC Supplier No. HYN22055MG
Esculentoside H (EsH) is a saponin isolated from the root extract of the perennial plant *Phytolacca esculenta*. It exhibits anti-tumor activity and suppresses colon cancer cell migration by blocking the JNK1/2 and NF-κB signaling-mediated matrix metalloproteinases-9 (MMP-9) expression, also linked to its capacity for TNF release.
  • Saponin isolated from the root extract of *Phytolacca esculenta*.
  • Demonstrates anti-tumor activity.
  • Inhibits colon cancer cell migration.
  • Blocks JNK1/2 and NF-κB signaling.
  • Suppresses matrix metalloproteinases-9 (MMP-9) expression.
  • Linked to TNF release.