Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Esculentoside H | 66656-92-6 | 98.7% | 989.10 g·mol⁻¹ | C48H76O21 | 10 MG
SDP

Catalog No. 5000296637
Encompass_Preferred

Esculentoside H is a triterpenoid saponin isolated from Phytolacca esculenta and reported as an NF-κB inhibitor with demonstrated anti-tumor activity. It is supplied as a white to off-white solid for research use in biochemical and cell-based studies.

  • High purity: 98.7%
  • CAS number: 66656-92-6
  • Molecular formula: C48H76O21
  • Molecular weight: 989.10 g·mol⁻¹
  • Solubility: soluble in DMSO (≈100 mg/mL; hygroscopic)
  • Storage: store at 4°C; protect from light; in solution store at -80°C (6 months) or -20°C (1 month)
  • Pack size: 10 mg

Catalog No. 50-002-96637 Supplier Medchemexpress LLC Supplier No. HYN220510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Esculentoside H is a triterpenoid saponin isolated from Phytolacca esculenta and reported as an NF-κB inhibitor with demonstrated anti-tumor activity. It is supplied as a white to off-white solid for research use in biochemical and cell-based studies.

  • High purity: 98.7%
  • CAS number: 66656-92-6
  • Molecular formula: C48H76O21
  • Molecular weight: 989.10 g·mol⁻¹
  • Solubility: soluble in DMSO (≈100 mg/mL; hygroscopic)
  • Storage: store at 4°C; protect from light; in solution store at -80°C (6 months) or -20°C (1 month)
  • Pack size: 10 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.