Learn More
Medchemexpress LLC Esculetin | 305-01-1 | 99.79% | 178.14 | 1 ML
SDP
Catalog No. 5000427220
Encompass_Preferred
Esculetin is an active ingredient extracted mainly from the bark of *Fraxinus rhynchophylla*. It primarily functions by inhibiting the PI3K/Akt pathway, which affects platelet-derived growth factor (PDGF)-induced airway smooth muscle cells (ASMCs) phenotype switching. Additionally, Esculetin demonstrates antioxidant, anti-inflammatory, and antitumor properties, making it a subject of various research applications. It is intended for research use only and not for medical or patient use.
  • Inhibits platelet-derived growth factor (PDGF)-induced airway smooth muscle cell (ASMC) proliferation and migration.
  • Demonstrates antioxidant activity.
  • Exhibits anti-inflammatory effects.
  • Possesses antitumor activities.
  • Targets PI3K pathway.
  • Induces apoptosis in certain cancer cell types.
  • Ameliorates skin lesions in psoriatic mice models.
Catalog No. 50-004-27220 Supplier Medchemexpress LLC Supplier No. HYN028410MM/1ML
Esculetin is an active ingredient extracted mainly from the bark of *Fraxinus rhynchophylla*. It primarily functions by inhibiting the PI3K/Akt pathway, which affects platelet-derived growth factor (PDGF)-induced airway smooth muscle cells (ASMCs) phenotype switching. Additionally, Esculetin demonstrates antioxidant, anti-inflammatory, and antitumor properties, making it a subject of various research applications. It is intended for research use only and not for medical or patient use.
  • Inhibits platelet-derived growth factor (PDGF)-induced airway smooth muscle cell (ASMC) proliferation and migration.
  • Demonstrates antioxidant activity.
  • Exhibits anti-inflammatory effects.
  • Possesses antitumor activities.
  • Targets PI3K pathway.
  • Induces apoptosis in certain cancer cell types.
  • Ameliorates skin lesions in psoriatic mice models.