Learn More
Selleck Chemical LLC Esculetin S4711-100mg
SDP
Catalog No. 503109563
Encompass_Preferred
Esculetin (Cichorigenin Aesculetin) is a coumarin derivative found in various natural plant products with various biological and pharmaceutical properties including anti-edema anti-inflammatory and anti-tumour effects It inhibits lipoxygenases (LOs)
Catalog No. 50-310-9563 Supplier Selleck Chemical LLC Supplier No. S4711100MG
Esculetin (Cichorigenin Aesculetin) is a coumarin derivative found in various natural plant products with various biological and pharmaceutical properties including anti-edema anti-inflammatory and anti-tumour effects It inhibits lipoxygenases (LOs)