Learn More
Selleck Chemical LLC Esculetin S4711-1g
SDP
Catalog No. 503109928
Encompass_Preferred
Esculetin (Cichorigenin Aesculetin) is a coumarin derivative found in various natural plant products with various biological and pharmaceutical properties including anti-edema anti-inflammatory and anti-tumour effects It inhibits lipoxygenases (LOs)
Catalog No. 50-310-9928 Supplier Selleck Chemical LLC Supplier No. S47111G
Esculetin (Cichorigenin Aesculetin) is a coumarin derivative found in various natural plant products with various biological and pharmaceutical properties including anti-edema anti-inflammatory and anti-tumour effects It inhibits lipoxygenases (LOs)