Learn More
Medchemexpress LLC HY-N0188 500mg Medchemexpress, Esculin CAS:531-75-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0188500MG
Encompass_Preferred
Medchemexpress, HY-N0188 500mg Esculin CAS:531-75-9 Esculin, a fluorescent coumarin glucoside, is an active ingredient of ash bark. Esculin ameliorates cognitive impairment in experimental diabetic nephropathy (DN), and exerts anti‑oxidative stress and anti‑inflammatory effects, via the MAPK signaling pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2777