Learn More
Apexbio Technology LLC Esculin 531-75-9 20mg
SDP
Catalog No. 5000089623
Encompass_Preferred
Esculin (CAS 531-75-9) is a coumarin glucoside isolated from the bark of Aesculus hippocastanum It functions primarily as a -glucosidase substrate hydrolyzing to esculetin and glucose upon enzymatic action Esculin s fluorescence properties make it valuable in assays monitoring -glucosidase activity in microbiological and biochemical research Additionally its antioxidant and vasoprotective effects have supported investigations into microcirculatory disorders and oxidative stress-related models Esculin is soluble in hot ethanol methanol pyridine ethyl acetate and acetic acid and should be stored in sealed cool and dry conditions
Catalog No. 50-000-89623 Supplier Apexbio Technology LLC Supplier No. N176620
Esculin (CAS 531-75-9) is a coumarin glucoside isolated from the bark of Aesculus hippocastanum It functions primarily as a -glucosidase substrate hydrolyzing to esculetin and glucose upon enzymatic action Esculin s fluorescence properties make it valuable in assays monitoring -glucosidase activity in microbiological and biochemical research Additionally its antioxidant and vasoprotective effects have supported investigations into microcirculatory disorders and oxidative stress-related models Esculin is soluble in hot ethanol methanol pyridine ethyl acetate and acetic acid and should be stored in sealed cool and dry conditions