Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Esculin monohydrate, 99.07%, MP Biomedicals™

Catalog No. ICN10165525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10165525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10165525 Supplier MP Biomedicals, Inc Supplier No. 0210165525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 531-75-9
Molecular Formula C15H16O9
Molecular Weight (g/mol) 340.28
MDL Number MFCD00149492
InChI Key XHCADAYNFIFUHF-TYKRLAFXNA-N
Synonym esculin, aesculin, esculoside, --esculin, polychrome, 6,7-dihydroxycoumarin-6-o-glucoside, aesculinum, esculine, esculetin 6-o-glucoside, 6,7-dihydroxycoumarin 6-glucoside
PubChem CID 5281417
ChEBI CHEBI:4853
IUPAC Name 7-hydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
SMILES OC[C@H]1O[C@@H](OC2=C(O)C=C3OC(=O)C=CC3=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Color White
Assay Percent Range 0.9907
Quantity 25 g
Formula Weight 358.28
Physical Form Crystalline Powder
Chemical Name or Material Esculin monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.