Learn More
Aviva Systems Biology Corp ESD Antibody - N-terminal region (ARP58619_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58619_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ESD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ESD. Alternate names: FGHApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62376