Learn More
MSC LPD ESD/Antistatic Bucket, 4 gal, Polypropylene, Static Dissipative, 1 ea
Catalog No. 502868289
Encompass
The LPD ESD/Antistatic Bucket is designed for use in environments where static electricity needs to be controlled. This round, single-pail bucket has a 4-gallon capacity and is constructed from static dissipative polypropylene, including the handle, to prevent electrostatic discharge.
  • Capacity: 4 gal
  • Polypropylene bucket material
  • Single pail style
  • Polypropylene handle material
  • Round shape
  • Black color
  • Handle included
  • Static dissipative
Catalog No. 50-286-8289 Supplier MSC Supplier No. 20447983
The LPD ESD/Antistatic Bucket is designed for use in environments where static electricity needs to be controlled. This round, single-pail bucket has a 4-gallon capacity and is constructed from static dissipative polypropylene, including the handle, to prevent electrostatic discharge.
  • Capacity: 4 gal
  • Polypropylene bucket material
  • Single pail style
  • Polypropylene handle material
  • Round shape
  • Black color
  • Handle included
  • Static dissipative