Learn More
Fisherbrand ESD Chair
Catalog No. 14360618 Shop All Fisherbrand Products
Change view
Click to view available options
Seating Type:
Desk Height
Catalog No. Seating Type
14-360-618 Desk Height
1 options
Catalog No. 14-360-618
Get 1 Extra Fisherbrand Chair When You Buy 4
View all promotions
Add to Cart
Add to Cart
With ESD casters
  • Adjustable from 17.5 to 22.75 in. from the top of the seat to the floor
  • Features seat height adjustment
  • Cast aluminum base
  • Electro-static dissipating, dual-wheel, non-marring casters (suitable for both hard and carpeted floors)
  • Chemical, soil, and puncture resistant polyurethane
  • Meets ANSI/ESD STM12.1-2013
  • Electro-static dissipating components to ensure proper grounding
Specifications
Product Type ESD Chair
Back Style Standard
With Armrest No
With Backrest Yes
Footrest No
With Lumbar Support No
With Tilt No
Color Blue
Autoclavable Not Autoclavable
With Casters Yes
Material Polyurethane
Seating Type Desk Height
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov