Learn More
Bevco Precision Manufacturing Co Versa Tall Height ESD/ISO 5 Cleanroom Black Vinyl Backless Stool; Polished Aluminum Base
Catalog No. NC2022945 Shop All Bevco Precision Manufacturing Co Products
Encompass_Preferred
BEVCO’s Versa ESD/ISO 5 Cleanroom Series models are designed to meet specific particulate goals of ISO 14644-1 and the ESD requirements of ANSI/ESD STM12. 1-2019 delivering a combination ESD/Cleanroom seating solution for highly technical environments. 16” diameter black vinyl seat that complies with CAL 117. The pneumatic height adjustment allows for stress-free seat height changes from 21. 5" - 31. 5". ESD black vinyl meets CAL 117 standards. The stool features a sturdy 25" diameter chrome tubular steel base with a brass drag chain. Model comes standard with ESD mushroom glides. Optional ESD casters can be ordered separately.
Catalog No. NC2022945 Supplier Bevco Precision Manufacturing Co Supplier No. 3550E2VBLK
BEVCO’s Versa ESD/ISO 5 Cleanroom Series models are designed to meet specific particulate goals of ISO 14644-1 and the ESD requirements of ANSI/ESD STM12. 1-2019 delivering a combination ESD/Cleanroom seating solution for highly technical environments. 16” diameter black vinyl seat that complies with CAL 117. The pneumatic height adjustment allows for stress-free seat height changes from 21. 5" - 31. 5". ESD black vinyl meets CAL 117 standards. The stool features a sturdy 25" diameter chrome tubular steel base with a brass drag chain. Model comes standard with ESD mushroom glides. Optional ESD casters can be ordered separately.