Learn More
Transforming Technologies™ ESD Jacket with Lapel Collar and Snap Cuff
SDP
Catalog No. 171273809
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-127-3809 5X-Large
17-127-3801 X-Small
17-127-3802 Small
17-127-3803 Medium
17-127-3804 Large
17-127-3805 X-Large
17-127-3806 2X-Large
17-127-3807 3X-Large
17-127-3808 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-127-3809 Supplier Transforming Technologies™ Supplier No. JKC9029SPWH
Only null left
Add to Cart
Add to Cart
9010 Series ESD Jackets with Lapel collar and knit cuff provide high-quality ESD protection are are fabricated from lightweight 90% polyester and 10% carbon fabric for unmatched operator comfort. These jackets feature a snap-front closure and three roomy patch pockets.
  • Medium weight ESD jacket ideal for all working environments and numerous applications
  • Made with highly durable materials and maintains consistent continuity readings, even after 100 wash cycles
  • Certified a Level 3 ESD Jacket (A Groundable Garment System)
Specifications
Size 5X-Large
Gender Unisex
Material Polyester
Color White
Closure Type Snap
Certifications/Compliance ANSI ESD S202.0
Collar Type Lapel
Cuff Style Snap
Disposable Reusable
Latex Free Yes
No. of Pockets 3
Resistance <1 x 106
Product Type ESD Jacket
Sleeve Style 3/4 Length