Learn More
CritiCore Protective Wear ESD Knit Full Length Lab Coats
Catalog No. 18111357
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-357 6X-Large
18-111-348 X-Small
18-111-349 Small
18-111-350 Medium
18-111-351 Large
18-111-352 X-Large
18-111-353 2X-Large
18-111-354 3X-Large
18-111-355 4X-Large
18-111-356 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 18-111-357 Supplier CritiCore Protective Wear Supplier No. 12L252FCEKC66XL01BLU
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lab coats offer protection against static build up and electrostatic discharge in a variety of industrial environments.
  • Bellton™ ESD knit cuff
  • Reusable ESD Knit fabric
  • Reinforced snap front closure
  • Grounding snaps on pockets and under arms
  • ESD compliance indicator label
  • Qualifies as a groundable, static control garment
Specifications
Product Type General Purpose Lab Coat
Size 6X-Large
Gender Unisex
Material Polyester
Color Blue
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Static
No. of Pockets 3
Collar Type Lapel
Latex Free Yes
Seam Type Pre-serged/Felled