Learn More
R&R Lotions ESD LAMIN SHEETS 8.5X11 100/PK
SDP
Supplier:  R&R Lotions SDSHEETS
Encompass_Preferred
Static dissipative lamination sheet works with any standard document laminating system. Laminate paper or signs and made them ESD Safe. 100 sheets per pack. Size: 8.5×11″.
Catalog No. NC0684176