Learn More
MSC LPD Trade ESD Conductive Lid, 15 L, Polypropylene, 1 ea
Catalog No. 502816855
Encompass
This ESD conductive lid is specifically designed for 15-liter buckets, providing an effective solution to safely dissipate static electricity and protect sensitive components.
  • Discharges static charge
  • Constructed from conductive polypropylene material
  • Compatible with 15 L anti-static buckets
  • Black color
Catalog No. 50-281-6855 Supplier MSC Supplier No. 20448023
This ESD conductive lid is specifically designed for 15-liter buckets, providing an effective solution to safely dissipate static electricity and protect sensitive components.
  • Discharges static charge
  • Constructed from conductive polypropylene material
  • Compatible with 15 L anti-static buckets
  • Black color