Learn More
ACL Inc ESD Microfiber Foam Swab, 2.75", 500 swabs per bag
Catalog No. NC1841788
Encompass_Preferred
ACL’s ESD Microfiber Foam Swab is an exceptional quality swab forthe critical cleaning of ESD static-sensitive surfaces. Constructed ofmedical-grade foam wrapped in microfiber knit, this swab is highlyeffective in removing and holding liquid or dry contaminants involume. The rigid ESD handle has a dissipative resistance and willnot generate a charge.The inner foam pad adds increased liquid holding capacity andcushion while microfiber enhances pick-up and can withstandabrasion. These characteristics make it ideal to use with solventssuch as acetone, ethanol, IPA, and methanol and for applying orremoving lubricants in hard-to-reach areas.
Catalog No. NC1841788 Supplier ACL Inc Supplier No. 7250
ACL’s ESD Microfiber Foam Swab is an exceptional quality swab forthe critical cleaning of ESD static-sensitive surfaces. Constructed ofmedical-grade foam wrapped in microfiber knit, this swab is highlyeffective in removing and holding liquid or dry contaminants involume. The rigid ESD handle has a dissipative resistance and willnot generate a charge.The inner foam pad adds increased liquid holding capacity andcushion while microfiber enhances pick-up and can withstandabrasion. These characteristics make it ideal to use with solventssuch as acetone, ethanol, IPA, and methanol and for applying orremoving lubricants in hard-to-reach areas.