Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro ESD Preconfigured Utility Cart

Catalog No. 01001351 Shop All Metro Products
Change view
Click to view available options
Dimensions (L x W x H):
42 x 18 in. 39 in.
48 x 24 x 39 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
01-001-351 48 x 24 x 39 in.
01-001-333 -
01-001-335 42 x 18 in. 39 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-001-351 Supplier Metro Supplier No. ESDMW2448NC2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metro Preconfigured Utility Cart, 2-tier wire.

  • 2-tier wire
  • They are not autoclave-friendly

Specifications

Autoclavable Not Autoclavable
CAS 67-56-1, 1346600-40-5
Color Silver
Dimensions (L x W x H) 48 x 24 x 39 in.
Load Bearing Capacity (English) 375 lb.
Load Bearing Capacity (Metric) 170 kg
Linear Formula C10H13ClFNO
Material Chrome
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
No. of Shelves 2
Formula Weight 217.67
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.