Learn More
Electron Microscopy Sciences EMS ESD Tweezers Rubber Coated Style 4, Anti-Acid/Anti-MagSS, 110 mm OAL
SDP
Catalog No. 501930600
Encompass_Preferred
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed. Straight, extra fine tips.
OAL: 110 mm
Tips: Anti-Acid/Anti-MagSS
Material: Straight, extra fine
Catalog No. 50-193-0600 Supplier Electron Microscopy Sciences Supplier No. 783254DR
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed. Straight, extra fine tips.
OAL: 110 mm
Tips: Anti-Acid/Anti-MagSS
Material: Straight, extra fine