Learn More
Electron Microscopy Sciences EMS ESD Tweezers Rubber Coated Style 5C, Anti-Acid/Anti-MagSS, 115 mm OAL
SDP
Catalog No. 501930603
Encompass_Preferred
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed. Extra fine, double bent tips.
OAL: 115 mm
Tips: Anti-Acid/Anti-MagSS
Material: Extra fine, double bent
Catalog No. 50-193-0603 Supplier Electron Microscopy Sciences Supplier No. 783255C
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed. Extra fine, double bent tips.
OAL: 115 mm
Tips: Anti-Acid/Anti-MagSS
Material: Extra fine, double bent