Learn More
MSC EMist ESD Safety Toe-Strap Connector, Grounding, Static Prevention, 1 ea
Catalog No. 502818689
Encompass
The EMist ESD Safety Toe-Strap Connector is a critical component of the grounding strap system designed to prevent static charge accumulation during the operation of the EMist EM360 Electrostatic Sprayer.
  • Essential for proper operation of the EM360 electrostatic sprayer system
  • Part of a grounding strap that prevents static charge accumulation
  • Features a simple loop that slips over the shoe's toe and heel
  • Includes a white portion for the ball of the shoe and an adjustable black elastic strap for the heel
Catalog No. 50-281-8689 Supplier MSC Supplier No. 10301562
The EMist ESD Safety Toe-Strap Connector is a critical component of the grounding strap system designed to prevent static charge accumulation during the operation of the EMist EM360 Electrostatic Sprayer.
  • Essential for proper operation of the EM360 electrostatic sprayer system
  • Part of a grounding strap that prevents static charge accumulation
  • Features a simple loop that slips over the shoe's toe and heel
  • Includes a white portion for the ball of the shoe and an adjustable black elastic strap for the heel