Learn More
MSC LPD ESD Anti Static Squeegee, 19.69 in, Straight, 1 ea
Catalog No. 502816854
Encompass
The LPD ESD Anti Static Squeegee (base only) features a 19.69 in wide, straight frame and is black. Its base is composed of high-quality conductive polypropylene, and the bristles are made of conductive nylon, providing an ESD-safe cleaning solution for environments where electrostatics must be controlled.
  • ESD Squeegee
  • Blade is 19.69 in wide
  • Straight frame
  • Black color
  • Static dissipative
  • Base of conductive polypropylene
  • Bristles of conductive nylon
  • Designed for use with ESD handle
Catalog No. 50-281-6854 Supplier MSC Supplier No. 20448064
The LPD ESD Anti Static Squeegee (base only) features a 19.69 in wide, straight frame and is black. Its base is composed of high-quality conductive polypropylene, and the bristles are made of conductive nylon, providing an ESD-safe cleaning solution for environments where electrostatics must be controlled.
  • ESD Squeegee
  • Blade is 19.69 in wide
  • Straight frame
  • Black color
  • Static dissipative
  • Base of conductive polypropylene
  • Bristles of conductive nylon
  • Designed for use with ESD handle