Learn More
Transforming Technologies ESD Washable Cleanroom Shoe Cover with Grounding Ribbon, White
SDP
Catalog No. 17142021
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-142-021 Medium
17-142-020 Large
17-142-019 X-Large
17-142-018 2X-Large
17-142-017 3X-Large
17-142-015 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-142-021 Supplier Transforming Technologies Supplier No. SC50BSWH03
Add to Cart
Add to Cart
ESD Washable Cleanroom Shoe Cover with Grounding Ribbon is made with ESD rubber soles and ideal for cleanrooms, labs, manufacturing facilities, and other sterile environments.
Our SC50BS cleanroom shoe covers are made with ESD rubber soles and ideal for cleanrooms, labs, manufacturing facilities, and other sterile environments. The ESD washable cleanroom shoe covers help protect the integrity of cleanroom environments and protects against ESD damage due to static. Our shoes covers converts non-ESD footwear in cleanroom applications to footwear that will provide a path-to-ground for static charges. The shoe cover meets the ANSI/ESD S20.20 required limit tested per ESD SP9.2 & ESD TR53 as suitable ESD footwear component in personnel grounding flooring/footwear system. Because they’re reusable, the covers can also save you time and money – no more need to order disposable shoe covers, shoe booties, or boot covers.
The SC50BS cleanroom shoe covers made with static dissipative cleanroom fabric, dissipative ESD rubber for the soles and conductive ground ribbons to make contact with the body. The ESD washable cleanroom shoe covers work well with most common shoes and boot but also work well when worn inside laundered cleanroom boot covers such as our TX4000 ESD Soft Sole Boots. The surged-seam inside shoe cover design features an elastic opening that fits most regular shoe styles with a secure fit. A full length non-woven conductive carbon grounding ribbon is added to the bottom of our ESD Shoe Covers to remove the static from personnel.
Specifications
Size Medium
Material Polyester
Latex Free Yes
Color White
Disposable Reusable
Sterility Non-sterile
Closure Type Elastic Top with Adjustable Snap
Sole Type Anti-static
Certifications/Compliance ANSI/ESD S20.20 ESD SP9.2 & TR53
Resistance Static
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 1000 (ISO 6)