Learn More
Expression Systems ESF 921 CELL CULTURE MEDIA 20L
SDP
Catalog No. 501656602
Encompass_Preferred
ESF 921™ Insect Cell Culture Medium is a complete serum-free, protein-free medium developed for robust cell growth, protein production and baculovirus vector production for a wide range of insect cells including Sf9, Sf21, Tni (High Five™), and Drosophila S2. ESF 921 contains L-Glutamine and Kolliphor P188 (Pluronic F68).This serum-free, protein-free insect cell culture medium provides unmatched cell growth and expression for a wide range of insect cells including Sf9, Sf21, Tni (High Five™) and Drosophila S2.• Sf9 cell densities of 15 x 106 cells/ml or greater• Produces high titer baculovirus vector yields of 5E8 to 3E9 infectious units/ml• Complete, ready-to-use, contains L-glutamine• Ideal for scale-up manufacturing• Works with a wide variety of baculovirus and host cell platforms• In stock for immediate shipment• Adapted cells availableDownload COA: www.expressionsystems.com. Kolliphor(R) is a registered trademark of BASF SE.
Catalog No. 50-165-6602 Supplier Expression Systems Supplier No. 9600120
Add to Cart
Add to Cart
ESF 921™ Insect Cell Culture Medium is a complete serum-free, protein-free medium developed for robust cell growth, protein production and baculovirus vector production for a wide range of insect cells including Sf9, Sf21, Tni (High Five™), and Drosophila S2. ESF 921 contains L-Glutamine and Kolliphor P188 (Pluronic F68).This serum-free, protein-free insect cell culture medium provides unmatched cell growth and expression for a wide range of insect cells including Sf9, Sf21, Tni (High Five™) and Drosophila S2.• Sf9 cell densities of 15 x 106 cells/ml or greater• Produces high titer baculovirus vector yields of 5E8 to 3E9 infectious units/ml• Complete, ready-to-use, contains L-glutamine• Ideal for scale-up manufacturing• Works with a wide variety of baculovirus and host cell platforms• In stock for immediate shipment• Adapted cells availableDownload COA: www.expressionsystems.com. Kolliphor(R) is a registered trademark of BASF SE.