Learn More
Expression Systems ESF AF Insect Cell Culture Medium, Animal Free
SDP
Catalog No. NC1142434
Encompass_Preferred
1L Bottle
Expression Systems ESF AF is an animal origin-free formulation for insect cell culture. It has been thoroughly tested on Sf9, Sf21, Tni (High Five™) and S2 cell lines and is comparable to ESF 921 for growth and expression. ESF AF is the logical choice for further manufacturing under cGMP/FDA regulatory compliance and has a Drug Master File with the US FDA.
• 100% animal component-free
• Sf9 cell densities of 20 x 106 cells/ml or greater
• Ideal for scale-up manufacturing
• Works with a wide variety of baculovirus and host cell platforms
• Optimized for applications such as production of sub-unit vaccines, virus-like particles and gene therapy vectors (rAAV all serotypes)
• Available in 1L bottles and media transfer bags of 10, 20 and 50L volumes
• Complete, ready-to-use, contains L-glutamine
Always in stock for immediate shipment
Download Instructions for Use and/or COA please visit: www.expressionsystems.com
Catalog No. NC1142434 Supplier Expression Systems Supplier No. 9930001
1L Bottle
Expression Systems ESF AF is an animal origin-free formulation for insect cell culture. It has been thoroughly tested on Sf9, Sf21, Tni (High Five™) and S2 cell lines and is comparable to ESF 921 for growth and expression. ESF AF is the logical choice for further manufacturing under cGMP/FDA regulatory compliance and has a Drug Master File with the US FDA.
• 100% animal component-free
• Sf9 cell densities of 20 x 106 cells/ml or greater
• Ideal for scale-up manufacturing
• Works with a wide variety of baculovirus and host cell platforms
• Optimized for applications such as production of sub-unit vaccines, virus-like particles and gene therapy vectors (rAAV all serotypes)
• Available in 1L bottles and media transfer bags of 10, 20 and 50L volumes
• Complete, ready-to-use, contains L-glutamine
Always in stock for immediate shipment
Download Instructions for Use and/or COA please visit: www.expressionsystems.com