Learn More
Expression Systems ESF SFM Serum Free Cell Culture Medium For CHO & 293 Cells, 1L Bottle
SDP
Catalog No. NC0572183
Encompass_Preferred
Expression Systems ESF SFM cell culture medium is a serum-free, animal-free formulation designed for robust growth and expression of HEK 293, CHO and hybridoma cells in suspension. It is a complete and ready-to-use medium, designed for use in shake flasks or bioreactors.
• Serum-free, animal-free medium for HEK 293, CHO and hybridoma cells
• Complete, ready-to-use, contains L-glutamine
• Optimized for robust cell growth and protein expression
• Available in 1L bottles
Appropriate for suspension and adherent cultures (contains surfactant)
Ideal for scale-up manufacturing
Always in stock for immediate shipment
• Adapted cells available in suspension
To download Instructions for Use and/or COA please visit: www.expressionsystems.com
Catalog No. NC0572183 Supplier Expression Systems Supplier No. 98001
Expression Systems ESF SFM cell culture medium is a serum-free, animal-free formulation designed for robust growth and expression of HEK 293, CHO and hybridoma cells in suspension. It is a complete and ready-to-use medium, designed for use in shake flasks or bioreactors.
• Serum-free, animal-free medium for HEK 293, CHO and hybridoma cells
• Complete, ready-to-use, contains L-glutamine
• Optimized for robust cell growth and protein expression
• Available in 1L bottles
Appropriate for suspension and adherent cultures (contains surfactant)
Ideal for scale-up manufacturing
Always in stock for immediate shipment
• Adapted cells available in suspension
To download Instructions for Use and/or COA please visit: www.expressionsystems.com