Learn More
Medchemexpress LLC Esfenvalerate | 66230-04-4 | 98% | 50 MG
SDP
Catalog No. 5000338400
Encompass_Preferred
Esfenvalerate is an analytical standard for research and analytical applications. It is one of the four isomers of the pyrethroid insecticide fenvalerate.
  • Analytical standard
  • Reference standard supplied assay
  • Used in qualitative, quantitative, and methodological research experiments
  • Applications include HPLC, GC, and MS
Catalog No. 50-003-38400 Supplier Medchemexpress LLC Supplier No. HY12925750MG
Esfenvalerate is an analytical standard for research and analytical applications. It is one of the four isomers of the pyrethroid insecticide fenvalerate.
  • Analytical standard
  • Reference standard supplied assay
  • Used in qualitative, quantitative, and methodological research experiments
  • Applications include HPLC, GC, and MS