Learn More
Medchemexpress LLC Esfenvalerate | 66230-04-4 | 99.7% | 1 ML
SDP
Catalog No. 5000342285
Encompass_Preferred
Esfenvalerate is one of the four isomers of fenvalerate, a pyrethroid insecticide. Intended for research and analytical applications only, studies have shown that Esfenvalerate at doses of 15 mg/kg/day (gavage, from gestation day 13 to 19) can exhibit clinical signs of neurotoxicity.
  • Suitable for research and analytical applications
  • Isomer of fenvalerate, a pyrethroid insecticide
  • Exhibits neurotoxicity in certain studies
Catalog No. 50-003-42285 Supplier Medchemexpress LLC Supplier No. HY12925710MM/1ML
Esfenvalerate is one of the four isomers of fenvalerate, a pyrethroid insecticide. Intended for research and analytical applications only, studies have shown that Esfenvalerate at doses of 15 mg/kg/day (gavage, from gestation day 13 to 19) can exhibit clinical signs of neurotoxicity.
  • Suitable for research and analytical applications
  • Isomer of fenvalerate, a pyrethroid insecticide
  • Exhibits neurotoxicity in certain studies