Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Esfenvalerate | 66230-04-4 | 99.7% | 100 MG
SDP

Catalog No. 5000340642
Encompass_Preferred

Esfenvalerate is one of four isomers of the pyrethroid insecticide fenvalerate, intended for research use only. It presents as a white to light yellow solid and has a molecular weight of 419.90. This compound is soluble in DMSO at 100 mg/mL.

  • For research use only
  • One of four isomers of pyrethroid insecticide fenvalerate
  • Appears as a white to light yellow solid
  • Soluble in DMSO at 100 mg/mL
  • Molecular formula C25H22ClNO3
  • Studies showed clinical signs of neurotoxicity at 15 mg/kg/day in vivo

Catalog No. 50-003-40642 Supplier Medchemexpress LLC Supplier No. HY129257100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Esfenvalerate is one of four isomers of the pyrethroid insecticide fenvalerate, intended for research use only. It presents as a white to light yellow solid and has a molecular weight of 419.90. This compound is soluble in DMSO at 100 mg/mL.

  • For research use only
  • One of four isomers of pyrethroid insecticide fenvalerate
  • Appears as a white to light yellow solid
  • Soluble in DMSO at 100 mg/mL
  • Molecular formula C25H22ClNO3
  • Studies showed clinical signs of neurotoxicity at 15 mg/kg/day in vivo

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.