Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific eShield™ Sterile Covers

Catalog No. 22375400 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
SLR Camera
Smart Phone
Tablet
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-375-400 Smart Phone
22-375-402 Tablet
22-375-404 SLR Camera
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-375-400
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Save time and money by accessing vital programs and data in real time, safely within a sterile field

  • Sterile cover for smart phones
  • Constructed of ultra-clear polyethylene for clear photos and is touchscreen compatible with gloves
  • Protects the sterile environment from contaminants on handheld devices
  • Easily accommodates aseptic technique

Recommended Storage

General Warehouse Storage - Keep out of direct sunlight

Specifications

For Use With (Equipment) Smart Phone
Material Polyethylene Film
Sterility Sterile
Dimensions (L x W) 22.9 x 35.6 cm (9 x 14 in.)
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.