Learn More
Medchemexpress LLC HY-117656 5mg Medchemexpress, ESI-05 CAS:5184-64-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1176565MG
Encompass_Preferred
Medchemexpress, HY-117656 5mg ESI-05 CAS:5184-64-5 ESI-05 (NSC 116966) is a specific antagonist of EPAC2 (exchange protein directly activated by cAMP 2), with an IC 50 of 0.4 µM. ESI-05 (NSC 116966) inhibits cAMP-mediated activation of EPAC2 as well as EAPC2 mediated Rap1 activation [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2526