Learn More
Medchemexpress LLC ESI-08 | 301177-43-5 | 98.7% | 353.48 | 1 ML
SDP
Catalog No. 5000351611
Encompass_Preferred
ESI-08 is a potent and selective EPAC antagonist. It completely inhibits both EPAC1 and EPAC2 activity, selectively blocking cAMP-induced EPAC activation without inhibiting cAMP-mediated PKA activation. Exchange proteins directly activated by cAMP (EPAC) are guanine nucleotide exchange factors regulating intracellular processes in response to the second messenger cAMP.
  • Potent and selective EPAC antagonist
  • Completely inhibits EPAC1 and EPAC2 activity
  • Selectively blocks cAMP-induced EPAC activation
  • Does not inhibit cAMP-mediated PKA activation
Catalog No. 50-003-51611 Supplier Medchemexpress LLC Supplier No. HY13617210MM/1ML
ESI-08 is a potent and selective EPAC antagonist. It completely inhibits both EPAC1 and EPAC2 activity, selectively blocking cAMP-induced EPAC activation without inhibiting cAMP-mediated PKA activation. Exchange proteins directly activated by cAMP (EPAC) are guanine nucleotide exchange factors regulating intracellular processes in response to the second messenger cAMP.
  • Potent and selective EPAC antagonist
  • Completely inhibits EPAC1 and EPAC2 activity
  • Selectively blocks cAMP-induced EPAC activation
  • Does not inhibit cAMP-mediated PKA activation