Learn More
TARGETMOL CHEMICALS INC ESI-09 25MG
SDP
Catalog No. 502580954
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ESI-09 is a new-typel noncyclic nucleotide EPAC antagonist.The IC50 values of ESI-09 for EPAC1 and EPAC2 is 3.2 and 1.4 uM respectively. purity: 99%
Catalog No. 50-258-0954 Supplier TARGETMOL CHEMICALS INC Supplier No. T224425MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ESI-09 is a new-typel noncyclic nucleotide EPAC antagonist.The IC50 values of ESI-09 for EPAC1 and EPAC2 is 3.2 and 1.4 uM respectively. purity: 99%