Learn More
Selleck Chemical LLC ESI-09 5mg
SDP
Supplier:  Selleck Chemical LLC S74995MG
Encompass_Preferred
ESI-09 is a specific exchange protein directly activated by cAMP (EPAC) inhibitor with IC50 of 3.2 ?M and 1.4 ?M for EPAC1 and EPAC2, respectively, >100-fold selectivity over PKA. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3279